Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipocalin-2/NGAL Rat anti-Mouse, Canine (Negative), Human (Negative), Porcine (Negative), Rat (Negative), Alexa Fluor 532, Clone: 10, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP221746AF532

Catalog No. NP221746A32


Add to cart

Description

Description

Lipocalin-2/NGAL Monoclonal antibody specifically detects Lipocalin-2/NGAL in Mouse, Canine (Negative), Human (Negative), Porcine (Negative), Rat (Negative) samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Lipocalin-2/NGAL
10
Western Blot, ELISA
Conjugated, Purified
LCN2
Recombinant human NGAL adsorbed onto aluminum hydroxide gel.
Protein A or G purified
Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuroscience
Primary
3934.0
Canine (Negative), Human (Negative), Mouse, Porcine (Negative), Rat (Negative)
ELISA, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin
Rat
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
NBP2-21746 binds NGAL in mouse serum.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit