Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lipocalin-2/NGAL Rat anti-Mouse, DyLight 755, Clone: 29, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP105183IR
Description
Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Lipocalin-2/NGAL | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LCN2 | |
Recombinant mouse NGAL adsorbed onto aluminum hydroxide gel | |
0.1 mL | |
Lipid and Metabolism | |
3934 | |
Mouse | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
29 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin | |
Rat | |
Protein A purified | |
RUO | |
Primary | |
neutrophil gelatinase-associated lipocalin | |
Store at 4C in the dark. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title