Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lipofectamine™ 2000 Transfection Reagent Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.

Lipofectamine™ 2000 Transfection Reagent

$424.00

Specifications

Final Product Type Lipofectamine™ 2000 Transfection Reagent
For Use With (Application) Transfection
Sample Type Plasmid DNA, RNAi Plasmids (shRNA, miR), Synthetic siRNA
Temperature (English) Operating 35.6 to 46.4°F
Temperature (Metric) Operating 2 to 8°C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-668-027 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only. Green Product View Documents Invitrogen™
11668027
0.75mL Each for $424.00
Add to cart
 
Description

Description

Lipofectamine™ 2000 Transfection Reagent is a versatile transfection reagent that has been shown to effectively transfect the widest variety of adherent and suspension cell lines. Researchers use Lipofectamine™ 2000 Reagent for siRNA- and shRNA-based gene knockdown experiments, as well as for gene expression studies. Lipofectamine™ 2000 Transfection Reagent is our best transfection reagent available for the co-transfection of siRNA and plasmid DNA.

With Lipofectamine™ 2000 Transfection Reagent, you'll get:

  • Exceptional transfection efficiency in the broadest range of cell lines and the highest levels of recombinant protein expression (see table)
  • Superior performance for co-transfection of siRNA and plasmid DNA
  • Proven efficacy in the presence of serum—eliminates the need to change media following transfection
  • Reliable performance for high-throughput applications
  • The best choice for establishing stable cell lines

    A high-performance transfection reagent for gene expression and gene silencing
    Lipofectamine™ 2000 Transfection Reagent works effectively with all common cell lines as well as with many challenging ones, and can be used in media with or without serum. For gene silencing, Lipofectamine™ 2000 Transfection Reagent's high-efficiency transfections provide the high levels of gene knockdown needed to achieve convincing results. Lipofectamine™ 2000 Transfection Reagent is the number one choice for co-transfection, given its effectiveness for transfecting both siRNA and plasmid DNA. Lipofectamine™ 2000 Transfection Reagent is easy to use—simply mix with nucleic acid and add to cell culture.

    Ideal for high-throughput work
    Simplicity and speed combined with high transfection efficiency make Lipofectamine™ 2000 Transfection Reagent ideal for transient protein expression or high-throughput RNAi transfections. Transfection conditions can be easily established for automated or robotic systems used in such applications.

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

  • Specifications

    Specifications

    Lipofectamine™ 2000 Transfection Reagent
    Plasmid DNA, RNAi Plasmids (shRNA, miR), Synthetic siRNA
    2 to 8°C
    Transfection
    35.6 to 46.4°F
    Lipofectamine™ 2000 Transfection Reagent

    For Research Use Only. Not for use in diagnostic procedures.

    Provide Content Correction

    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit