Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lipofectamine™ RNAiMAX Transfection Reagent Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.

Lipofectamine™ RNAiMAX Transfection Reagent

$225.00 - $6840.00

Specifications

Final Product Type Lipofectamine™ RNAiMAX Transfection Reagent
For Use With (Application) RNAi, transfection
Sample Type Synthetic siRNA
Temperature (English) Operating 35.6 to 46.4°F
Temperature (Metric) Operating 2 to 8°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
13-778-030 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only. View Documents Invitrogen™
13778030
0.3mL Each for $225.00
Add to cart
 
13-778-500 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only. View Documents Invitrogen™
13778500
15mL Each for $6,840.00
Add to cart
 
Description

Description

Lipofectamine™ RNAiMAX Transfection Reagent provides the highest transfection efficiencies on the widest variety of cell types for siRNA-mediated gene knockdown experiments. Lipofectamine™ RNAiMAX is a proprietary RNAi-specific cationic lipid formulation designed specifically for the delivery of siRNA and miRNA into all cell types.

With Lipofectamine™ RNAiMAX Transfection Reagent you will get:

  • Superior transfection efficiency requiring lower RNAi concentrations, leading to more effective gene knockdown with minimal nonspecific effects
  • Easy optimization due to minimal cytotoxicity across a 10-fold concentration range of transfection reagent
  • Superior transfection efficiencies for miRNA antagonists and mimics
  • Compatibility with a broad range of cell types, providing the most versatile approach to all of your gene silencing experiments
  • A simple and rapid protocol for consistent and reproducible results

    High knockdown in a wide range of cells
    Lipofectamine™ RNAiMAX Transfection Reagent transfects a wide range of cell types (see figure). For gene silencing, Lipofectamine™ RNAiMAX Transfection Reagent's high-efficiency transfections lead to the high levels of gene knockdown needed to achieve convincing results.

    Simple, high-throughput–ready transfections
    Simply mix Lipofectamine™ RNAiMAX Transfection Reagent with siRNA, add to your cells, incubate, and measure gene knockdown. The simplicity and speed combined with high transfection efficiency make Lipofectamine™ RNAiMAX Transfection Reagent ideal for high-throughput siRNA transfections. Transfection conditions can be easily established for automated or robotic systems used in such applications.

    Find an optimized Lipofectamine™ RNAiMAX transfection protocol for your cell line.

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

  • Specifications

    Specifications

    Lipofectamine™ RNAiMAX Transfection Reagent
    Synthetic siRNA
    2 to 8°C
    RNAi, transfection
    35.6 to 46.4°F
    Lipofectamine™ RNAiMAX Transfection Reagent
    Safety and Handling

    Safety and Handling

    ShelfLife : 6 Month(s)

    For Research Use Only. Not for use in diagnostic procedures.

    Provide Content Correction

    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit