Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipolysis Stimulated Lipoprotein Receptor Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Lipolysis Stimulated Lipoprotein Receptor
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094117
View Documents
Novus Biologicals
NBP303749100UL
100 μg
Each for $320.71
Add to cart
 
NB094118
View Documents
Novus Biologicals
NBP30374920UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

Lipolysis Stimulated Lipoprotein Receptor Polyclonal antibody specifically detects Lipolysis Stimulated Lipoprotein Receptor in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

Lipolysis Stimulated Lipoprotein Receptor
Unconjugated
Rabbit
Human, Mouse, Rat
ILDR3, immunoglobulin-like domain containing receptor 3, lipolysis stimulated lipoprotein receptor, lipolysis-stimulated lipoprotein receptor, lipolysis-stimulated receptor, lipolysis-stimulated remnant, LISCH, LISCH protein, LISCH7, liver-specific bHLH-Zip transcription factor, MGC10659, MGC48312, MGC48503
Recombinant fusion protein containing a sequence corresponding to amino acids 50-240 of human Lipolysis Stimulated Lipoprotein Receptor (NP_057009.3).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
51599
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit