Learn More
Diazyme Laboratories Lipoprotein (a) Control
Diazyme Lipoprotein (a) Control set is for the testing of Lipoprotein (a) in samples of human serum.
Manufacturer:  Diazyme Laboratories DZ131BCON
Catalog No. 23-700-544
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
  • A dual vial, liquid stable reagent system which provides reliable results
  • The Lp(a) assay features time saving instrument specific packaging options which eliminate reagent transfer and increases operator efficiency
  • The assay method requires only 6 μL of serum sample and is completed in less than 10 minutes
  • Has a correlation with existing commercial products with a correlation coefficient of 0.9868, slope of 0.9828, and y intercept of 1.0033
  • Lp(a)'s linear range extends up to 100 mg/dL and has Within-Run and Between-Run precision CV values ≤ 3.7%
Specifications
Specifications
LIPOPROTEIN (A) CONTROL SET, 2 LV (lyophilized)
Lipoprotein (a) [Lp(a)]
The Diazyme Lipoprotein(a) Control Set is a two-level set that is supplied in liquid stable form (2 x 1 mL). The CONTROL is manu-factured from human serum. The concentration of the CONTROL is lot specific and the Lipoprotein(a) concentrations are expressed in mg/dL.
Store at 2 to 8°C. DO NOT FREEZE. Note: Store CONTROL tightly capped when not in use.
Diagnostic
2 x 1 mL vial
Unopened vials: stability is up to the expiration date on label when stored at 2 to 8°C. Opened Vials: open vials are stable up to 45 days when stored at 2 to 8°C.
CE, 510(k) Cleared
Consists of 2 vials