Learn More
MP Biomedicals™ Lipoxidase, MP Biomedicals™
Catalyzes the dioxygenation of polyunsaturated fatty acids. Lipoxidase, MP Biomedicals is an iron-containing enzyme that catalyse the hydroperoxidation of lipids containing a cis,cis-1,4- pentadiene structure. Products of lipoxygenases are involved in div
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP21988136 View Documents MP Biomedicals™
02198813.6
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MP21988132 View Documents MP Biomedicals™
02198813.2
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Lipoxygenases are found in plants, animals and fungi. The soybean enzyme will use arachidonic acid as a substrate, with ∼15% of the activity indicated using linoleic acid as the substrate; the product of arachidonic acid oxidation is 12- or 15-hydroperoxyarachidonic acid (12-HPETE or 15-HPETE).