Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LIPT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559119
Description
Immunogen sequence: CAIGVRCGRH ITSHGLALNC STDLTWFEHI VPCGLVGTGV TSLSKELQRH VTVEEVMPPF LVAFKEIYKC TLISEDS Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
Catalyzes the transfer of endogenously produced octanoic acid from octanoyl-acyl-carrier-protein onto the lipoyl domains of lipoate-dependent enzymes. Lipoyl-ACP can also act as a substrate although octanoyl-ACP is likely to be the physiological substrate.
Specifications
LIPT2 | |
Polyclonal | |
Unconjugated | |
LIPT2 | |
Lipoate-protein ligase B; lipoyl(octanoyl) transferase 2 (putative); lipoyl/octanoyl transferase; LIPT2; Octanoyl-[acyl-carrier-protein]-protein N-octanoyltransferase; putative lipoyltransferase 2, mitochondrial; putative octanoyltransferase, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
387787 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NK58 | |
LIPT2 | |
Recombinant protein corresponding to Human LIPT2. Recombinant protein control fragment (Product #RP-97493). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction