Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Liquefied Phenol, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
2.5 L
Specifications
Specifications
| Quantity | 2.5 L |
| Specific Gravity | 1.05 |
| Physical Form | Liquid |
| For Use With (Application) | Disinfectant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction