Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scientific Cell Company Liquid Filter Set for Testing Photometric Accuracy in UV Range

For testing photometric accuracy (0.75 Abs) in the UV range, according to European Pharmacopoeia.

Supplier:  Scientific Cell Company 667.301UV

Catalog No. 14-397-521


Explore available promotions
Add to Cart

Includes: UV60: Absorption filter, UV14: Reference filter, DAkkS calibration certificates

Description

Description

  • Certified at: wavelengths: 235, 257, 313, 350nm
  • Slit width: 2nm
  • Supplied in high-quality storage case
Specifications

Specifications

Liquid Filter Set
Testing photometric accuracy (0.75 Abs) in the UV range, according to European Pharmacopoeia
UV60: Absorption filter, UV14: Reference filter, DAkkS calibration certificates
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.