Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Scientific Cell Company Liquid Filter Set for Testing Wavelength Accuracy
Shop All Scientific Cell Company Inc. Products
Click to view available options
Type:
Liquid Filter Set
Description
Includes
UV5: Wavelength filter, UV14: Reference filter, DAkkS calibration certificates
- Certified at wavelengths: 241, 287, 361, 536, 640nm
- Slit width: 1nm
- Supplied in high-quality storage case
Specifications
Specifications
| Type | Liquid Filter Set |
| For Use With (Application) | Testing wavelength accuracy, according to European Pharmacopoeia |
| Includes | UV5: Wavelength filter, UV14: Reference filter, DAkkS calibration certificates |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction