Learn More
BioTek™ Liquid Handling Control Software for MultiFlo™ Microplate Dispenser - PROMO
Offers dual mode control - runs protocols from PC or instrument keypad
Manufacturer:  BioTek™ LHC2PM
Catalog No. 11-120-675PM
Add to cart
This item is not returnable. View return policy
Description
Description
Full PC control for BioTek MultiFlo Microplate Dispenser with assay-specific protocol requirement definition in a familiar Windows™ environment.
  • Intuitive StepWise™ protocol creation with an unlimited number of methods
  • Create and run protocols from PC or download to instrument's keypad
  • Automatic protocol validation for error-free assurance
  • Protocol parameters printed for safe record keeping and GLP compliance
  • Custom maintenance reminders for continued instrument performance in busy laboratories
  • Supports BioStack integrations for high throughput requirements
  • Security enabled protection for downloaded protocols
  • Multiple instruments controlled in parallel
Specifications
Specifications
Liquid Handling Control Software
1/Case
Documents
Promotions
Promotions
  • BioTek InstrumentsSupplier Diversity Partner
    Get 50% off BioTek Gen5 and other Liquid Handling Control Software.
    06-30-2019