Learn More
MetaSystems™ Liquillizer™ Mucolytic Agent
Liquillizer™ Mucolytic Reagent is ideal for fast, efficient, user friendly preparation of clinical sputum and other respiratory samples for microbiology or mycobacteriology testing.
Manufacturer:  MetaSystems™ P7510002IG
 View more versions of this product
Catalog No. 22-200-371
Add to cart
Description
Description
  • Ready to Use, No Preparation or Mixing
  • Odorless and DTT Free
  • Room Temprature Storage
  • No Incubation, Centrifuging, or Sample Heating
  • 4 Week Open Container Shelf Life
Specifications
Specifications
Non-sterile
Diagnostic procedures and cultivation
1
250 mL
Storage and use at room temperature (15 to 25°C)
Safety and Handling
Safety and Handling
ShelfLife : 4 weeks after opening
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Liquillizer™ is intended for the liquefaction of viscous respiratory specimens such as sputa, tracheal secretions, bronchial secretions or bronchoalveolar lavage. The mucolytic Dithiothreitol (DTT) -free buffer reduces specimen viscosity for easier sample handling while maintaining organism viability. No interference with molecular methods has been observed.