Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (±)-Lisofylline, 98%

Click to view available options
Quantity:
10mg
25mg
50mg
6493-06-7
C13H20N4O3
280.33
MFCD00871850
NSMXQKNUPPXBRG-UHFFFAOYNA-N
+/--lisofylline,hydroxy pentoxifylline,lisofylline usan:inn,1-r-5-hydroxyhexyl theobromine,lisophylline,1-5'-hydroxyhexyl-3,7-dimethylxanthine,r-lisophylline,3,7-dimethyl-1-5-hydroxyhexyl xanthine,1-5r-hydroxyhexyl-3,7-dimethylxanthine,xanthine, 1-5'-hydroxyhexyl-3,7-dimethyl
57782
1-(5-hydroxyhexyl)-3,7-dimethylpurine-2,6-dione
CC(CCCCN1C(=O)C2=C(N=CN2C)N(C1=O)C)O
6493-06-7
C13H20N4O3
280.33
MFCD00871850
NSMXQKNUPPXBRG-UHFFFAOYNA-N
+/--lisofylline,hydroxy pentoxifylline,lisofylline usan:inn,1-r-5-hydroxyhexyl theobromine,lisophylline,1-5'-hydroxyhexyl-3,7-dimethylxanthine,r-lisophylline,3,7-dimethyl-1-5-hydroxyhexyl xanthine,1-5r-hydroxyhexyl-3,7-dimethylxanthine,xanthine, 1-5'-hydroxyhexyl-3,7-dimethyl
57782
1-(5-hydroxyhexyl)-3,7-dimethylpurine-2,6-dione
CC(CCCCN1C(=O)C2=C(N=CN2C)N(C1=O)C)O
Quantity 25mg
Formula Weight 280.33
Physical Form Solid
Percent Purity 98%
Chemical Name or Material (±)-Lisofylline