Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ (±)-Lisofylline, 98%
Click to view available options
Quantity:
10mg
25mg
50mg
Specifications
Quantity | 50mg |
Formula Weight | 280.33 |
Physical Form | Solid |
Percent Purity | 98% |
Chemical Name or Material | (±)-Lisofylline |