Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (±)-Lisofylline, 98%

Click to view available options
Quantity:
10mg
25mg
50mg
6530-09-2
C7H16Cl2N2
199.12
MFCD00871850
STZHBULOYDCZET-UHFFFAOYNA-N
3-aminoquinuclidine dihydrochloride,1-azabicyclo 2.2.2 octan-3-amine dihydrochloride,quinuclidine, 3-amino-, dihydrochloride,3aminoquinuclidine dihydrochloride,quinuclidin-3-amine dihydrochloride,1-aza-bicyclo 2.2.2 oct-3-ylamine dihydrochloride,1-azabicyclo 2.2.2 octan-3-amine, dihydrochloride,3-amino-1-azabicyclo 2.2.2 octane dihydrochloride,quinuclidine-3-ylamine, chloride, chloride,1-azabicyclo 2.2.2 octan-3-amine, dihydrochloride
197853
1-azabicyclo[2.2.2]octan-3-amine;dihydrochloride
C1CN2CCC1C(C2)N.Cl.Cl
6530-09-2
C7H16Cl2N2
199.12
MFCD00871850
STZHBULOYDCZET-UHFFFAOYNA-N
3-aminoquinuclidine dihydrochloride,1-azabicyclo 2.2.2 octan-3-amine dihydrochloride,quinuclidine, 3-amino-, dihydrochloride,3aminoquinuclidine dihydrochloride,quinuclidin-3-amine dihydrochloride,1-aza-bicyclo 2.2.2 oct-3-ylamine dihydrochloride,1-azabicyclo 2.2.2 octan-3-amine, dihydrochloride,3-amino-1-azabicyclo 2.2.2 octane dihydrochloride,quinuclidine-3-ylamine, chloride, chloride,1-azabicyclo 2.2.2 octan-3-amine, dihydrochloride
197853
1-azabicyclo[2.2.2]octan-3-amine;dihydrochloride
C1CN2CCC1C(C2)N.Cl.Cl
Quantity 10mg
Formula Weight 280.33
Physical Form Solid
Percent Purity 98%
Chemical Name or Material (±)-Lisofylline