Learn More
Aat BioquestSupplier Diversity Partner Lissamine Rhodamine B Sulfonyl Chloride [Sulforhodamine B sulfonyl chloride] *CAS 62796-29-6* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 470
SRB sulfonyl chloride (also called Lissamine rhodamine sulfonyl chloride™) is highly fluorescent. It is quite unstable in water, especially at the higher pH required for reaction with aliphatic amines.
Catalog No. 50-195-2891