Learn More
Listeria flagella Mouse anti-Bacteria, Clone: 4220, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA533275
Catalog No. PIMA533275
Description
Description
Listeria is a genus of bacteria. Until 1992, 10 species were known, each containing two subspecies. By 2019, 20 species had been identified. The genus received its current name, after the British pioneer of sterile surgery Joseph Lister, in 1940. Listeria species are Gram-positive, rod-shaped, and facultatively anaerobic, possess flagella and do not produce endospores. The major human pathogen in the genus Listeria is L. monocytogenes. It is usually the causative agent of the relatively rare bacterial disease listeriosis, an infection caused by eating food contaminated with the bacteria.
Specifications
Specifications
Listeria flagella
Monoclonal
100 μg/mL
PBS with 0.1% sodium azide; pH 7.2
Mouse
Protein A
RUO
Antibody
Monoclonal
ELISA, Immunocytochemistry, Immunofluorescence
4220
Unconjugated
Liquid
IgG2b
1 mL
4° C
Primary
Bacteria