Learn More
Microbiologics™ Listeria innocua derived from ATCC 33090
EZ-FPC lyophilized QC microorganism preparations are daily process controls for both qualitative and quantitative detection methods. They’re ideal for Statistical Process Charting programs to standardize quality controls throughout your laboratory.
Manufacturer:  Microbiologics™ 0814FPC
Catalog No. 10-810-327
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
  • Biosafety Level: 1
  • 10 pellets of a quantitative microorganism manufactured as a food process control.
  • Qualitative Preparations at a concentration of 1.0-9.9E+02 CFU per pellet; serotype 6a
Specifications
Specifications
EZ-FPC
ATCC 33090
EZ-FPC
US
FDA Bacteriological Analytical Manual (BAM), ISO 11133
Plastic Canister
1
Food and Food Ingredients: Pathogen Testing, Media Testing
10 Pellets
Safety and Handling
Safety and Handling
ShelfLife : 8 Months (Minimum)