Learn More
Microbiologics™ Listeria innocua derived from NCTC 11288, Epower
Epower is the embodiment of flexibility and versatility. Use one product for all of your QC needs – from media QC and presence/absence testing to enumeration testing, water testing, and disinfectant testing. Use a single strain or combine multiple strains for a mixed population.
Manufacturer: Microbiologics™ 0414E3
This item is not returnable.
View return policy
Description
- Biosafety Level: 1
- 10 pellets of a single microorganism with a predetermined quantitative assay
- Serotype 6a
- Mean Assay Value: 1.0-9.9E+03 CFU per pellet
Specifications
NCTC 11288 | |
ISO 11133 | |
Epower | |
Plastic Canister | |
10 pellets | |
2 to 8°C |
1 | |
Food and Food Ingredients: Pathogen Testing, Media Testing | |
United States | |
Epower | |
8 Months (Minimum) |