Learn More
Microbiologics™ Listeria innocua derived from NCTC 11288, LYFO DISK
SDP
Simply rehydrate LYFO DISK qualitative microorganism pellets with a diluent and you’re ready to inoculate your culture media. LYFO DISK, packaged in a glass vial containing 6 pellets of a single strain, offers a ton of flexibility.
Supplier:  Microbiologics™ 0414L
Catalog No. 23-047-293
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
Biosafety Level: 16 lyophilized pellets of a single microorganism strain for quality controlserotype 6a
Specifications
Specifications
NCTC 11288
ISO 11133
LYFO DISK
Lyfo-Disk QC Control
Glass Vial
1
2°C to 8°C
United States
Media Testing, Microbial Identification
6 Pellets
Safety and Handling
Safety and Handling
ShelfLife : 8 Months (Minimum)