Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Listeria monocytogenes antigen 2 Mouse anti-Bacteria, Clone: , Novus Biologicals™

Mouse Monoclonal Antibody

$343.85

Specifications

Antigen Listeria monocytogenes antigen 2
Clone 2:13 C5
Host Species Mouse
Isotype IgG1 Kappa
Target Species Bacteria
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP254650 View Documents Novus Biologicals
NBP254650
0.1 mg Each for $343.85
Add to cart
 
NBP254651 View Documents Novus Biologicals
NBP254651
0.1 mg Each for $343.85
Add to cart
 
Description

Description

Listeria monocytogenes antigen 2 Monoclonal antibody specifically detects Listeria monocytogenes antigen 2 in Bacteria samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Listeria monocytogenes antigen 2
Mouse
Bacteria
RUO
Purified
Primary
PBS with 0.5% Sodium Azide
2:13 C5
IgG1 Kappa
ELISA, Western Blot
Unconjugated
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
1.0 mg/mL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit