Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Listeria monocytogenes p60, Mouse anti-Bacteria, Clone: p6007, Novus Biologicals™
Mouse Monoclonal Antibody
$374.00 - $542.50
Specifications
Antigen | Listeria monocytogenes p60 |
---|---|
Clone | p6007 |
Concentration | 1 mg/mL |
Dilution | Western Blot, ELISA |
Applications | Western Blot, ELISA |
Description
Listeria monocytogenes p60 Monoclonal specifically detects Listeria monocytogenes p60 in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
Listeria monocytogenes p60 | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Bacteria | |
IAP p60 from Listeria Monocytogenes, Invasion-associated Protein p60, Invasion-associated Protein p60 from Listeria Monocytogenes, invasion-associated-protein, p60 protein | |
IgG1 κ | |
Protein G purified | |
Recognizes Listeria monocytogenes p60 protein. Does not cross-react with other Listeria species. |
p6007 | |
Western Blot, ELISA | |
Monoclonal | |
Purified | |
RUO | |
0.2um-filtered solution in PBS, pH 7.4. | |
Recombinant Listeria monocytogenes p60. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title