Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Listeria monocytogenes p60, Mouse anti-Bacteria, Clone: p6017, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP280121100UG

 View more versions of this product

Catalog No. NB013247


Add to Cart

Description

Description

Listeria monocytogenes p60 Monoclonal specifically detects Listeria monocytogenes p60 in Bacteria samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Listeria monocytogenes p60
Monoclonal
1 mg/mL
Western Blot, ELISA
IAP p60 from Listeria Monocytogenes, Invasion-associated Protein p60, Invasion-associated Protein p60 from Listeria Monocytogenes, invasion-associated-protein, p60 protein
Recombinant Listeria spp. p60.
100 μg
Primary
Bacteria
Purified
Western Blot, ELISA
p6017
Unconjugated
0.2um-filtered solution in PBS, pH 7.4.
Mouse
Protein G purified
RUO
RecognizesListeria monocytogenes p60 protein. Weakly cross-reacts with other Listeria species (L. innocua, L. ivanovii).
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1 κ
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit