Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Listeria monocytogenes p60, Mouse anti-Bacteria, Clone: p6017, Novus Biologicals™

Mouse Monoclonal Antibody

$261.50 - $398.00

Specifications

Antigen Listeria monocytogenes p60
Clone p6017
Applications Western Blot, ELISA
Host Species Mouse
Target Species Bacteria
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB013247
View Documents
Novus Biologicals
NBP280121100UG
100 ug
Each for $398.00
Add to cart
 
NB013248
View Documents
Novus Biologicals
NBP28012150UG
50 ug
Each for $261.50
Add to cart
 
Description

Description

Listeria monocytogenes p60 Monoclonal antibody specifically detects Listeria monocytogenes p60 in Bacteria Samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Listeria monocytogenes p60
Western Blot, ELISA
Bacteria
Primary
p6017
Mouse
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit