Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Listeria Monocytogenes Rabbit anti-Mouse, Bacteria, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB100656670.1ML
Description
Listeria Monocytogenes Polyclonal specifically detects Listeria Monocytogenes in Mouse, Bacteria samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Listeria Monocytogenes | |
Polyclonal | |
Unconjugated | |
Phosphate buffered saline with 0.1% Sodium Azide | |
Rabbit | |
IgG purified | |
RUO | |
Reacts with Listeria monocytogenes whole cells. This product has not been absorbed and may react with related micro-organisms. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
4.0 mg/mL | |
ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
L. monocytogenes | |
Listeria monocytogenes; ATCC strain 43251 | |
0.1 mL | |
Primary | |
Bacteria, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only