Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Listeria Monocytogenes Rabbit anti-Mouse, Bacteria, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
$176.85 - $436.00
Specifications
Antigen | Listeria Monocytogenes |
---|---|
Concentration | 4.0 mg/mL |
Dilution | ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10065667
|
Novus Biologicals
NB10065667 |
0.5 mL |
Each for $436.00
|
|
NB017740
|
Novus Biologicals
NB100656670.1ML |
0.1 mL |
Each for $176.85
|
|
Description
Listeria Monocytogenes Polyclonal specifically detects Listeria Monocytogenes in Mouse, Bacteria samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Listeria Monocytogenes | |
ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Polyclonal | |
Purified | |
RUO | |
Phosphate buffered saline with 0.1% Sodium Azide | |
Listeria monocytogenes; ATCC strain 43251 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
4.0 mg/mL | |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Bacteria | |
L. monocytogenes | |
IgG | |
IgG purified | |
Reacts with Listeria monocytogenes whole cells. This product has not been absorbed and may react with related micro-organisms. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title