Alfa Aesar™ Lithium 2,4-pentanedionate, 99.5%

Manufacturer: Alfa Aesar™ 3052909

Quantity 10g

 View more versions of this product

Catalog No. AA3052909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18115-70-3
    Formula Weight 106.05
    MDL Number MFCD00013496
    Beilstein 4575087
    Synonym Lithium acetylacetonate
    Chemical Name or Material Lithium 2,4-pentanedionate

    Specifications

    Melting Point ≈250°C (dec.)
    Formula Weight 106.05
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym Lithium acetylacetonate
    Beilstein 4575087
    Health Hazard 1 Danger
    Color White
    Sensitivity Hygroscopic
    Physical Form Crystalline
    MDL Number MFCD00013496
    CAS 18115-70-3
    Quantity 10g
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Lithium 2,4-pentanedionate
    Assay 99.5%

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit