Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium 3,5-diiodosalicylate, MilliporeSigma™ Supelco™

$31.60 - $383.50

Specifications

Grade Analytical Standard
Linear Formula C7H3I2 LiO3
Physical Form Neat
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-5078
View Documents
MilliporeSigma Supelco
D36355G
5 g
Each for $104.20
Add to cart
 
11-101-5077
View Documents
MilliporeSigma Supelco
D363525G
25 g
Each for $383.50
Add to cart
 
11-101-5079
View Documents
MilliporeSigma Supelco
D36351G
1 g
Each for $31.60
Add to cart
 
Description

Description

Specifications

Specifications

Analytical Standard
C7H3I2 LiO3
Neat
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit