Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium acetylide ethylenediamine complex, 85%, ACROS Organics™

Click to view available options
Quantity:
50g
100g
6867-30-7
C2HLi·C2H8N2
92.07
MFCD00013183
WMWSRIHFAVOHSW-UHFFFAOYSA-N
lithium acetylide ethylenediamine complex,lithium acetylide, ethylenediamine complex,lithium; ethane-1,2-diamine; ethyne,1,2-ethanediamine compd. with lithium acetylide li c2h,lithium, 1,2-ethanediamine-kn1,kn2 ethynyl,1,2-ethanediamine, compd. with lithium acetylide li c2h 1:?,c2h8n2.c2hli,lithium acetylide, compd. with 1,2-ethanediamine,lithium acetylide, ethylene diamine complex 1:1,lithium acetylide, compound with ethane-1,2-diamine
2724010
lithium;ethane-1,2-diamine;ethyne
[Li+].C#[C-].C(CN)N
6867-30-7
C2HLi·C2H8N2
92.07
MFCD00013183
WMWSRIHFAVOHSW-UHFFFAOYSA-N
lithium acetylide ethylenediamine complex,lithium acetylide, ethylenediamine complex,lithium; ethane-1,2-diamine; ethyne,1,2-ethanediamine compd. with lithium acetylide li c2h,lithium, 1,2-ethanediamine-kn1,kn2 ethynyl,1,2-ethanediamine, compd. with lithium acetylide li c2h 1:?,c2h8n2.c2hli,lithium acetylide, compd. with 1,2-ethanediamine,lithium acetylide, ethylene diamine complex 1:1,lithium acetylide, compound with ethane-1,2-diamine
2724010
lithium;ethane-1,2-diamine;ethyne
[Li+].C#[C-].C(CN)N
Alkylation reagent
Quantity 50g
Melting Point 76.0°C
Color Yellow
Boiling Point 110.6°C
Assay Percent Range 80% min. (Gasvolumetric)
Linear Formula HC≡CLi·H2NCH2CH2NH2
Fieser 01,574; 05,382; 07,195; 11,288; 01,95; 02,51; 04,60; 05,78; 06,85; 07,45; 08,65; 09,83; 10,68; 11,101; 12,96; 13,56; 14,63; 15,59; 16,55; 17,59
Solubility Information Solubility in water: vigorous reaction. Other solubilities: soluble in ethylene diamine,n-propylamine and,pyridine
Formula Weight 92.07
Physical Form Crystals or Crystalline Powder
Percent Purity 85%
Chemical Name or Material Lithium acetylide ethylenediamine complex, 85%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

GHS P Statement
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Brush off loose particles from skin.
Immerse in cool water/wrap in wet bandages.
IF SWAL

GHS Signal Word: Danger

EINECSNumber : 229-967-9

TSCA : TSCA