Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium aluminium hydride, 15% solution in toluene/THF, ACROS Organics™

Click to view available options
Quantity:
800mL
Packaging:
AcroSeal™ Glass Bottle
16853-85-3,108-88-3,109-99-9
AlH4Li
37.954
MFCD00011075
BJKLPLABXHXMIM-UHFFFAOYSA-N
lithium aluminum hydride,lithium aluminium hydride,aluminum lithium hydride,lithiumaluminiumhydride,aluminum iii lithium hydride,lithium alanate,lithium aluminum tetrahydride,lithiumaluminiumhydrid,litiumaluminum hydride,lithim aluminum hydride
21226445
aluminum;lithium;hydride
[H-].[H-].[H-].[H-].[Li+].[Al+3]
16853-85-3,108-88-3,109-99-9
AlH4Li
37.954
MFCD00011075
BJKLPLABXHXMIM-UHFFFAOYSA-N
lithium aluminum hydride,lithium aluminium hydride,aluminum lithium hydride,lithiumaluminiumhydride,aluminum iii lithium hydride,lithium alanate,lithium aluminum tetrahydride,lithiumaluminiumhydrid,litiumaluminum hydride,lithim aluminum hydride
21226445
aluminum;lithium;hydride
[H-].[H-].[H-].[H-].[Li+].[Al+3]
Reduction reagent
Appearance of Solution Turbid solution
Linear Formula LiAlH4
Concentration ca 15%, exact strength on the certificate of analysis
Density 0.8900g/mL
Formula Weight 37.95
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.89
Quantity 800mL
Flash Point −20°C
Chemical Name or Material Lithium aluminium hydride, 15% solution in toluene/THF

GHS H Statement
Causes severe skin burns and eye damage. Suspected of damaging the unborn child. May be fatal if swallowed and enters airways. May cause damage to organs through prolonged or repeated exposure. May cause drowsiness or dizziness. May cause respiratory irritation. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. May form explosive peroxides. Suspected of causing cancer.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger