Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium aluminium hydride, 4.0M solution in diethyl ether, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
16853-85-3, 60-29-7
AlH4Li
37.954
MFCD00011075
BJKLPLABXHXMIM-UHFFFAOYSA-N
lithium aluminum hydride, lithium aluminium hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
21226445
aluminum;lithium;hydride
[H-].[H-].[H-].[H-].[Li+].[Al+3]
16853-85-3, 60-29-7
AlH4Li
37.954
MFCD00011075
BJKLPLABXHXMIM-UHFFFAOYSA-N
lithium aluminum hydride, lithium aluminium hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
21226445
aluminum;lithium;hydride
[H-].[H-].[H-].[H-].[Li+].[Al+3]
Reduction reagent
Linear Formula LiAIH4
Fieser 01,581; 02,242; 03,176; 04,291; 05,382; 06,325; 07,196; 08,286; 09,274; 10,236; 11,289; 12,272; 13,61; 14,190; 15,184; 16,133; 17,162
Merck Index 15, 344
Density 0.7100g/mL
Formula Weight 37.95
Physical Form Liquid
Percent Purity 18.0 to 22.0% (as LiAlH4)
Packaging AcroSeal™ Glass bottle
Solubility Solubility in water: vigorous reaction.
Specific Gravity 0.71
Storage Information Product may develop some turbidity or precipitate
Color Colorless
Quantity 100mL
Flash Point −017°C
Chemical Name or Material Lithium aluminium hydride, 4.0M Solution in Diethyl Ether
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage. Harmful if swallowed. May cause drowsiness or dizziness. In contact with water releases flammable gases which may ignite spontaneously. Extremely flammable liquid and vapor. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Keep away from any possible contact with water, because of violent reaction and possible flash fire. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray.

Danger