Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium aluminium hydride, 95%, powder, ACROS Organics™

Click to view available options
Quantity:
10 g
25 g
100 g
500 g
16853-85-3
AlH4Li
37.954
MFCD00011075
BJKLPLABXHXMIM-UHFFFAOYSA-N
lithium aluminum hydride,lithium aluminium hydride,aluminum lithium hydride,lithiumaluminiumhydride,aluminum iii lithium hydride,lithium alanate,lithium aluminum tetrahydride,lithiumaluminiumhydrid,litiumaluminum hydride,lithim aluminum hydride
21226445
aluminum;lithium;hydride
[H-].[H-].[H-].[H-].[Li+].[Al+3]
16853-85-3
AlH4Li
37.954
MFCD00011075
BJKLPLABXHXMIM-UHFFFAOYSA-N
lithium aluminum hydride,lithium aluminium hydride,aluminum lithium hydride,lithiumaluminiumhydride,aluminum iii lithium hydride,lithium alanate,lithium aluminum tetrahydride,lithiumaluminiumhydrid,litiumaluminum hydride,lithim aluminum hydride
21226445
aluminum;lithium;hydride
[H-].[H-].[H-].[H-].[Li+].[Al+3]
Reduction reagent
Quantity 25 g
Density .9g/mL
Assay Percent Range 95%
Linear Formula LiAlH4
Fieser 01,581; 02,242; 03,176; 04,291; 05,382; 06,325; 07,196; 08,286; 09,274; 10,236; 11,289; 12,272; 13,61; 14,190; 15,184; 16,133; 17,162
Merck Index 15, 344
Solubility Information Solubility in water: reacts. Other solubilities: soluble in diethyl ether (39g/100g at 25 °C), thf/, toluene (70:30) (17g/100g at 25 °C), in thf/mtbe (, 25:75) (12g/100g at 25 °C), in thf/toluene (15:85), (5g/100g at 25 °C);, practically insoluble in hyd
Formula Weight 37.95
Physical Form Powder
Percent Purity 95%
Melting Point 125.0°C
Chemical Name or Material Lithium aluminium hydride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger