Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium aluminium hydride bis(tetrahydrofuran), 1M solution in toluene, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
123439-81-6,108-88-3
C8H20AlLiO2
182.17
123439-81-6,108-88-3
C8H20AlLiO2
182.17
Reduction reagent
Solubility Information Solubility in water: reacts
Formula Weight 182.17
Physical Form Liquid
Concentration 0.95 to 1.10M
Density 0.8750g/mL
Packaging AcroSeal™ Glass Bottle
Solubility Solubility in water: reacts.
Specific Gravity 0.875
Color Colorless
Quantity 100mL
Flash Point 7°C
Chemical Name or Material Lithium aluminium hydride bis(tetrahydrofuran)
Show More Show Less

GHS H Statement
May be harmful if swallowed and enters airways.
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Harmful in conta

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear prote

GHS Signal Word: Danger