Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lithium aluminum hydride, 10% in THF

Reducing agent for pharmaceuticals and organic chemicals

Manufacturer:  Alfa Aesar™ 45502AP

Quantity 500mL

 View more versions of this product

Catalog No. AA45502AP


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

16853-85-3
LiAlH4
MFCD00011075
14,349
H4AlLi
37.95
UN3399
Lithium aluminum hydride

Specifications

16853-85-3
H4AlLi
37.95
UN3399
10% in THF
Characteristic
Pale gray
Lithium aluminum hydride
Liquid
LiAlH4
MFCD00011075
14,349
0.90g/mL
Air & Moisture Sensitive
500mL

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H260-H314-H351-H302-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Suspected of causing cancer.

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit