Lithium aluminum hydride, 95% min, Alfa Aesar™

Reducing Agent For Pharmaceuticals, Perfumes, and Fine organic Chemicals; Source of Hydrogen

Manufacturer: Alfa Aesar 13442A1

Quantity 1kg

 View more versions of this product

Catalog No. AA13442A1

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 16853-85-3
    Physical Form Powder
    Assay Percent Range 95% min.
    Molecular Formula LiAlH4
    Formula Weight 37.95
    MDL Number MFCD00011075
    Synonym LAH
    Chemical Name or Material Lithium aluminum hydride

    Specifications

    Physical Form Powder
    Formula Weight 37.95
    Quantity 1kg
    Color Light gray
    DOT Information DOT Class: 4.3, Packaging Group: I
    EINECS Number 240-877-9
    Odor Odorless
    Assay Percent Range 95% min.
    Molecular Formula LiAlH4
    Appearance Light gray powder
    MDL Number MFCD00011075
    CAS 16853-85-3
    Synonym LAH
    Health Hazard 1 H260-H301-H314-H318: In contact with water releases flammable gases which may ignite spontaneously. Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage.
    Chemical Name or Material Lithium aluminum hydride

    H260-H301-H314-H318: In contact with water releases flammable gases which may ignite spontaneously. Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit