Learn More
Lithium aluminum hydride, 95% min, Thermo Scientific Chemicals
Description
Reducing agent for pharmaceuticals, perfumes and fine organic chemicals; source of hydrogen, propellant, and catalyst in polymerizationsLithium aluminum hydride is used as a reducing agent in various synthetic organic chemistry reactions. For example, it is used in the conversion of esters, carboxylic acids, acyl chlorides, aldehydes and ketones into their corresponding alcohols. It is also involved in the conversion of amide, nitro, nitrile, imine, oxime and azide compounds into their amines as well as reduces alkyl halides to alkanes. It reacts with sodium chloride to prepare sodium hydride. It acts as a potential hydrogen storage medium, which could be used for future fuel cell-powered vehicles. It is mainly used in the preparation of other tetrahydridoaluminumates like sodium aluminum hydride, potassium aluminum hydride and magnesium alanate. Furthermore, it has been used to prepare thermoplastic polyester polyamides from oleic acid and lithium-polymer batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >125°C (decomposition) |
Color | Gray |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 95% min. |
Solubility Information | Decomposes in water. Soluble in ether,diethyl ether and terahydrofuran. |
Formula Weight | 37.95 |
Odor | Odorless |
Sensitivity | Air and moisture sensitive |
Appearance | Light gray powder |
Show More |
Safety and Handling
Hazard Category | H260-H314-H500 |
Precautionary Statement | P223-P231+P232-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c |
DOTInformation | DOT Class: 4.3, Packaging Group: I |
EINECSNumber | 240-877-9 |
RTECSNumber | BD0100000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.