Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium amide, 95%, ACROS Organics™

Click to view available options
Quantity:
500g
Packaging:
Glass bottle
7782-89-0
H2LiN
22.963
MFCD00011093
AFRJJFRNGGLMDW-UHFFFAOYSA-N
lithium amide,lithamide,lithium amide li nh2,lithiumamide,unii-7393omu9lk,lithium azanide,hsdb 649,linh2,lithium amide 25g,lithium amide, powder
24532
lithium;azanide
[Li+].[NH2-]
7782-89-0
H2LiN
22.963
MFCD00011093
AFRJJFRNGGLMDW-UHFFFAOYSA-N
lithium amide,lithamide,lithium amide li nh2,lithiumamide,unii-7393omu9lk,lithium azanide,hsdb 649,linh2,lithium amide 25g,lithium amide, powder
24532
lithium;azanide
[Li+].[NH2-]
Color White to Gray,White through Gray
Assay Percent Range 95%
Linear Formula LiNH2
Fieser 01,600; 03,178; 05,392; 13,159; 17,311
Merck Index 15, 5578
Formula Weight 22.95
Physical Form Fine Powder
Percent Purity ≥94% (LiNH2)
Density 1.1700g/mL
Packaging Glass bottle
Quantity 500g
Chemical Name or Material Lithium amide, 95%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.
Reacts violently with water.
May form explosive peroxides.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 231-968-4

TSCA : TSCA