Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Bis(trifluoromethanesulfonimide), 99%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
90076-65-6
C2F6LiNO4S2
287.09
MFCD00210017
QSZMZKBZAYQGRS-UHFFFAOYSA-N
lithium bis trifluoromethanesulfonyl imide,lithium bis trifluoromethyl sulfonyl amide,lithium bis trifluoromethanesulphonyl imide,litfsi,lithium bis trifluoromethylsulfonyl azanide,lithium bis trifluoromethylsulfonyl imide,bis trifluoromethane sulfonimide lithium salt,bistrifluoromethanesulfonimide lithium salt,lithium bis trifluoromethanesulfonyl amide,lithium triflimide
3816071
lithium;bis(trifluoromethylsulfonyl)azanide
[Li+].C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
90076-65-6
C2F6LiNO4S2
287.09
MFCD00210017
QSZMZKBZAYQGRS-UHFFFAOYSA-N
lithium bis trifluoromethanesulfonyl imide,lithium bis trifluoromethyl sulfonyl amide,lithium bis trifluoromethanesulphonyl imide,litfsi,lithium bis trifluoromethylsulfonyl azanide,lithium bis trifluoromethylsulfonyl imide,bis trifluoromethane sulfonimide lithium salt,bistrifluoromethanesulfonimide lithium salt,lithium bis trifluoromethanesulfonyl amide,lithium triflimide
3816071
lithium;bis(trifluoromethylsulfonyl)azanide
[Li+].C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Melting Point 234°C to 238°C
Color White
Quantity 10g
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 99%
Solubility Information Solubility in water: 10mg/mL (20°C)
Formula Weight 287.09
Physical Form Hygroscopic Powder
Percent Purity 99%
Chemical Name or Material Lithium bis(trifluoromethanesulfonimide)
Show More Show Less

GHS H Statement
Harmful to aquatic life with long lasting effects.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.<

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rins

GHS Signal Word: Danger