Learn More
Alfa Aesar™ Lithium bis(trimethylsilyl)amide, stab. with hexane
Manufacturer:  Alfa Aesar™ 4584614
Quantity 25g
 View more versions of this product
Catalog No. AA4584614
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
4039-32-1
[(CH3)3Si]2NLi
MFCD00008261
8136861
Lithium bis (trimethylsilyl)amide, Stabilized with hexane
C6H18LiNSi2
167.33
UN2925
LHMDS; Lithium hexamethyldisilazide
Specifications
4039-32-1
C6H18LiNSi2
167.33
UN2925
LHMDS; Lithium hexamethyldisilazide
25g
Lithium bis (trimethylsilyl)amide, Stabilized with hexane
Damp Powder
[(CH3)3Si]2NLi
MFCD00008261
8136861
Off-white
Air & Moisture Sensitive
Safety and Handling
SDS & Safety Documents
GHS H Statement
H228-H314-H361
Flammable solid.
Causes severe skin burns and eye damage.
Suspected of damaging fertility or the unborn child.
GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
TSCA : TSCA
storageNote1 :
  • Packaged under Argon