Alfa Aesar™ Lithium bis(trimethylsilyl)amide, stab. with hexane

Manufacturer: Alfa Aesar™ 4584606

Quantity 5g

 View more versions of this product

Catalog No. AA4584606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4039-32-1
    Molecular Formula C6H18LiNSi2
    Linear Formula [(CH3)3Si]2NLi
    Formula Weight 167.33
    MDL Number MFCD00008261
    UN Number UN2925
    Beilstein 8136861
    Synonym LHMDS; Lithium hexamethyldisilazide
    Chemical Name or Material Lithium bis (trimethylsilyl)amide, Stabilized with hexane

    Specifications

    Physical Form Damp Powder
    Formula Weight 167.33
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 4039-32-1
    UN Number UN2925
    Synonym LHMDS; Lithium hexamethyldisilazide
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H228-H314-H361
    Flammable solid.
    Causes severe skin burns and eye damage.
    Suspected of damaging fertility or the unborn child.
    TSCA TSCA
    Molecular Formula C6H18LiNSi2
    Linear Formula [(CH3)3Si]2NLi
    MDL Number MFCD00008261
    Beilstein 8136861
    Color Off-white
    Quantity 5g
    Sensitivity Air & Moisture Sensitive
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
    Storage Note 1
    • Packaged under Argon
    Chemical Name or Material Lithium bis (trimethylsilyl)amide, Stabilized with hexane

    GHS H Statement
    H228-H314-H361
    Flammable solid.
    Causes severe skin burns and eye damage.
    Suspected of damaging fertility or the unborn child.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit