Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium bromide, 99.999%, (trace metal basis), extra pure, ACROS Organics™

Click to view available options
Quantity:
25g
250g
Packaging:
Glass bottle
7550-35-8
BrLi
86.84
MFCD00011077
AMXOYNBUYSYVKV-UHFFFAOYSA-M
lithium bromide,lithium monobromide,lithium bromide libr,lithiumbromide,libr,lithium 1+ ion bromide,lithium bromide, anhydrous,lithium bromide, ultra dry,bromolithium,lithium-bromide
82050
CHEBI:63042
lithium;bromide
[Li+].[Br-]
7550-35-8
BrLi
86.84
MFCD00011077
AMXOYNBUYSYVKV-UHFFFAOYSA-M
lithium bromide,lithium monobromide,lithium bromide libr,lithiumbromide,libr,lithium 1+ ion bromide,lithium bromide, anhydrous,lithium bromide, ultra dry,bromolithium,lithium-bromide
82050
CHEBI:63042
lithium;bromide
[Li+].[Br-]
For ultra trace analysis
Assay Percent Range 99.999%
Linear Formula LiBr
Fieser 01,604; 02,245; 04,297
Merck Index 15, 5582
Solubility Information Solubility in water: 1.49 kg/l (20°C). Other solubilities: slightly soluble in alcohol,ether,methanol,,acetone and glycol,slightly soluble in pyridine,soluble in amyl alcohol
Formula Weight 86.84
Physical Form Powder and Chunks
Percent Purity 99.999%
Grade Trace Metal Basis
Density 3.4600g/mL
Packaging Glass bottle
Solubility Solubility in water: 1.49 kg/l (20°C). Other solubilities: slightly soluble in alcohol, ether, methanol,, acetone and glycol, slightly soluble in pyridine, soluble in amyl alcohol
Specific Gravity 3.46
Total Trace Metal Impurities 10ppm max.
Boiling Point 1265.0°C
Color White
Melting Point 550.0°C
Quantity 25g
Chemical Name or Material Lithium bromide, Extra Pure
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protect

GHS Signal Word: Warning

EINECSNumber : 231-439-8