Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium bromide, 99%, pure, anhydrous, Acros Organics

Click to view available options
Quantity:
250g
1kg
2.5kg
Packaging:
Plastic Bottle
7550-35-8
BrLi
86.84
AMXOYNBUYSYVKV-UHFFFAOYSA-M
lithium bromide,lithium monobromide,lithium bromide libr,lithiumbromide,libr,lithium 1+ ion bromide,lithium bromide, anhydrous,lithium bromide, ultra dry,bromolithium,lithium-bromide
82050
CHEBI:63042
lithium;bromide
[Li+].[Br-]
7550-35-8
BrLi
86.84
AMXOYNBUYSYVKV-UHFFFAOYSA-M
lithium bromide,lithium monobromide,lithium bromide libr,lithiumbromide,libr,lithium 1+ ion bromide,lithium bromide, anhydrous,lithium bromide, ultra dry,bromolithium,lithium-bromide
82050
CHEBI:63042
lithium;bromide
[Li+].[Br-]
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula LiBr
Merck Index 15,5582
Solubility Information Solubility in water: 1.49 kg/l (20°C). Other solubilities: slightly soluble in alcohol,ether,methanol,,acetone and glycol,slightly soluble in pyridine,soluble in amyl alcohol
Formula Weight 86.84
Percent Purity 99%
Grade Pure
Density 3.4600g/mL
Packaging Plastic Bottle
Solubility Solubility in water: 1.49 kg/l (20°C). Other solubilities: slightly soluble in alcohol, ether, methanol,, acetone and glycol, slightly soluble in pyridine, soluble in amyl alcohol
Specific Gravity 3.46
Boiling Point 1265.0°C
Melting Point 550.0°C
pH 6 to 12 (4 % aq. soln.)
Quantity 2.5kg
Chemical Name or Material Lithium bromide
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protect

GHS Signal Word: Warning

EINECSNumber : 231-439-8