Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium bromide, 99+%, for analysis, anhydrous, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
10kg
7550-35-8
BrLi
86.84
MFCD00011077
AMXOYNBUYSYVKV-UHFFFAOYSA-M
lithium bromide,lithium monobromide,lithium bromide libr,lithiumbromide,libr,lithium 1+ ion bromide,lithium bromide, anhydrous,lithium bromide, ultra dry,bromolithium,lithium-bromide
82050
CHEBI:63042
lithium;bromide
[Li+].[Br-]
7550-35-8
BrLi
86.84
MFCD00011077
AMXOYNBUYSYVKV-UHFFFAOYSA-M
lithium bromide,lithium monobromide,lithium bromide libr,lithiumbromide,libr,lithium 1+ ion bromide,lithium bromide, anhydrous,lithium bromide, ultra dry,bromolithium,lithium-bromide
82050
CHEBI:63042
lithium;bromide
[Li+].[Br-]
Assay Percent Range 99% min. (Argentometry)
Linear Formula LiBr
Fieser 01,604; 02,245; 04,297
Merck Index 15,5582
Solubility Information Solubility in water: 1.49 kg/l (20°C). Other solubilities: slightly soluble in alcohol,ether,methanol,,acetone and glycol,slightly soluble in pyridine,soluble in amyl alcohol
Formula Weight 86.84
Physical Form Powder
Percent Purity 99+%
Grade Analysis
Calcium (Ca) 50ppm max.
Carbonate 300ppm max.
Chloride 1500ppm max.
Chlorine (Cl) 1500ppm max.
Copper (Cu) 3ppm max.
Density 3.4600g/mL
Insoluble in Water 0.005% max.
Insoluble Matter 0.005% max. (in water)
Iron (Fe) 10ppm max.
Loss on Drying 0.5% max. (400°C)
Magnesium (Mg) 20ppm max.
Potassium (K) 300ppm max.
Sodium (Na) 400ppm max.
Solubility Solubility in water: 1.49 kg/l (20°C). Other solubilities: slightly soluble in alcohol, ether, methanol,, acetone and glycol, slightly soluble in pyridine, soluble in amyl alcohol
Specific Gravity 3.46
Sulfate 200ppm max.
Boiling Point 1265.0°C
Color Gray to Black
Melting Point 550.0°C
pH 6 to 12 (4% soln. at 20°C)
Quantity 10kg
Chemical Name or Material Lithium bromide, 99+%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protect

GHS Signal Word: Warning

EINECSNumber : 231-439-8

RTECSNumber : OJ5755000

TSCA : TSCA