Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium carbonate, 98+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
Packaging:
Plastic bottle
554-13-2
CLi2O3
73.88
XGZVUEUWXADBQD-UHFFFAOYSA-L
lithium carbonate,dilithium carbonate,lithonate,lithobid,lithane,eskalith,lithotabs,carbonic acid lithium salt,liskonum,lithizine
11125
CHEBI:6504
dilithium;carbonate
[Li+].[Li+].C(=O)([O-])[O-]
554-13-2
CLi2O3
73.88
XGZVUEUWXADBQD-UHFFFAOYSA-L
lithium carbonate,dilithium carbonate,lithonate,lithobid,lithane,eskalith,lithotabs,carbonic acid lithium salt,liskonum,lithizine
11125
CHEBI:6504
dilithium;carbonate
[Li+].[Li+].C(=O)([O-])[O-]
For analysis
Assay Percent Range 98% min
Linear Formula Li2CO3
Merck Index 15,5583
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: soluble in diluted acids,practically insoluble in alcohol
Formula Weight 73.88
Physical Form Fine Crystalline Powder
Percent Purity 98+%
Grade Analysis
Chloride 0.005% max
Heavy Metals (as Pb) 0.005% max.
Insoluble Matter 0.05% max. (in HCl)
Iron (Fe) 0.005% max.
Nitrate 0.005% max.
Packaging Plastic bottle
Sodium (Na) 0.1% max.
Solubility Solubility in water: 13g/L (20°C). Other solubilities: soluble in diluted acids, practically insoluble in alcohol
Sulfate 0.3% max
Melting Point 720.0°C
Quantity 250g
Chemical Name or Material Lithium carbonate
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Warning

EINECSNumber : 209-062-5