Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium carbonate, 99%, pure, ACROS Organics™

Click to view available options
Quantity:
500g
2.5kg
10kg
Packaging:
Plastic bottle
Plastic drum
554-13-2
CLi2O3
73.88
MFCD00011084
XGZVUEUWXADBQD-UHFFFAOYSA-L
lithium carbonate,dilithium carbonate,lithonate,lithobid,lithane,eskalith,lithotabs,carbonic acid lithium salt,liskonum,lithizine
11125
CHEBI:6504
dilithium;carbonate
[Li+].[Li+].C(=O)([O-])[O-]
554-13-2
CLi2O3
73.88
MFCD00011084
XGZVUEUWXADBQD-UHFFFAOYSA-L
lithium carbonate,dilithium carbonate,lithonate,lithobid,lithane,eskalith,lithotabs,carbonic acid lithium salt,liskonum,lithizine
11125
CHEBI:6504
dilithium;carbonate
[Li+].[Li+].C(=O)([O-])[O-]
Assay Percent Range 99%
Linear Formula Li2CO3
Fieser 01,606; 03,183; 04,192; 05,396
Merck Index 15,5583
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: soluble in diluted acids,practically insoluble in alcohol
Formula Weight 73.88
Physical Form Finely Granulated Powder
Percent Purity 99%
Grade Pure
Density 2.11
Loss on Drying 0.4% max. (200°C)
Packaging Plastic bottle
Solubility Solubility in water: 13g/L (20°C). Other solubilities: soluble in diluted acids, practically insoluble in alcohol
Color White
Melting Point 720.0°C
Quantity 500g
Chemical Name or Material Lithium carbonate, 99%
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Warning

EINECSNumber : 209-062-5

RTECSNumber : OJ5800000

TSCA : TSCA