Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lithium chloride, 200mM aq. soln.

Click to view available options
Quantity:
100mL
250mL
7447-41-8
ClLi
42.39
MFCD00011078
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium chloride,lithium chloride licl,lithiumchloride,licl,chlorure de lithium,chlorku litu,chlorolithium,lithiumchlorid,chlorku litu polish,luthium chloride
433294
CHEBI:48607
lithium;chloride
[Li+].[Cl-]
7447-41-8
ClLi
42.39
MFCD00011078
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium chloride,lithium chloride licl,lithiumchloride,licl,chlorure de lithium,chlorku litu,chlorolithium,lithiumchlorid,chlorku litu polish,luthium chloride
433294
CHEBI:48607
lithium;chloride
[Li+].[Cl-]
Applications
Used in formation of an active Mn(0) species useful for radical cyclization reactions. It is utilized in a variety of assays to study cell-fate and neurobiology. In developing Xenopus embryos, lithium chloride is observed to exert inhibition of GSK-3β (glycogen synthase kinase-3β). Additionally, in transfected Chinese Hamster Ovary cells (CHO) stimulated with angiotension II (Ang II), it was noted that lithium chloride enhanced the production of inositol triphosphate. The antiviral properties of lithium chloride were noted in a study which showed that the compound inhibited pseudorabis virus infection in vitro. It is primarily used in the synthesis of lithium metal by electrolysis. Also acts as a prescribed drug for bipolar mood disorder, it may also exhibit anti-inflammatory properties.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol, ether, pyridine, nitrobenzene, acetone, and amyl alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Linear Formula LiCl
Formula Weight 42.39
Physical Form Liquid
Concentration 200mM aq. soln.
Odor Odorless
Color Colorless
Quantity 100mL
Chemical Name or Material Lithium chloride