Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium chloride, 99%, For Analysis, Anhydrous, Acros Organics™

Click to view available options
Quantity:
100g
500g
2kg
10kg
Packaging:
Plastic bottle
Plastic drum
7447-41-8
ClLi
42.38
MFCD00011078
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium chloride,lithium chloride licl,lithiumchloride,licl,chlorure de lithium,chlorku litu,chlorolithium,lithiumchlorid,chlorku litu polish,luthium chloride
433294
CHEBI:48607
lithium;chloride
[Li+].[Cl-]
7447-41-8
ClLi
42.38
MFCD00011078
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium chloride,lithium chloride licl,lithiumchloride,licl,chlorure de lithium,chlorku litu,chlorolithium,lithiumchlorid,chlorku litu polish,luthium chloride
433294
CHEBI:48607
lithium;chloride
[Li+].[Cl-]
Additional Information carbonate (as Li2CO3): < 0.03% (typically)
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula LiCl
Fieser 01,609; 02,246; 04,298; 05,677; 12,277; 16,194
Merck Index 15,5584
Solubility Information Solubility in water: 832g/L (20°C). Other solubilities: soluble in alcohols,ether,pyridine,,nitrobenzene and acetone
Barium (Ba) 200ppm max.
Formula Weight 42.38
Physical Form Crystalline Granules
Percent Purity 99%
Grade Analysis
Calcium (Ca) 100ppm max.
Density 2.0600g/mL
Insoluble in Water 0.005% max. (typically)
Insoluble Matter 0.005% max. (in HCl)
Loss on Drying 0.5% max. (400°C, 4 hrs)
Packaging Plastic bottle
Potassium (K) 300ppm max.
Sodium (Na) 2500ppm max.
Solubility Solubility in water: 832g/L (20°C). Other solubilities: soluble in alcohols, ether, pyridine,, nitrobenzene and acetone
Specific Gravity 2.06
Sulfate 0.01% max
Trace Analysis Trace analysis are typical values
Boiling Point 1382.0°C
Color White
Melting Point 605.0°C
Quantity 100g
Chemical Name or Material Lithium chloride, 99%
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse

GHS Signal Word: Warning

EINECSNumber : 231-212-3

RTECSNumber : OJ5950000

TSCA : TSCA