Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Chloride ACS AR Granular, Macron Fine Chemicals™

Click to view available options
Quantity:
125g
500g
2.5kg
Packaging:
POLYSTORMOR
Poly
7447-41-8
ClLi
42.39
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
433294
CHEBI:48607
lithium;chloride
[Li+].[Cl-]
7447-41-8
ClLi
42.39
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
433294
CHEBI:48607
lithium;chloride
[Li+].[Cl-]
Formula Weight 42.39
Physical Form Granular
Grade ACS
Packaging POLYSTORMOR
Quantity 125g
Chemical Name or Material Lithium Chloride