Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Chloride, Crystal, Reagent, ACS, 99%, Spectrum™

Click to view available options
Quantity:
12 kg
125 g
125 lbs.
2.5 kg
25 g
500 g
6 x 125 g
6 x 500 g
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7447-41-8
ClLi
42.39
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium(1+) chloride
[Li+].[Cl-]
7447-41-8
ClLi
42.39
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium(1+) chloride
[Li+].[Cl-]

L1109, 7447-41-8, LiCl

Spectrum™ Lithium Chloride, Crystal, Reagent, ACS is a chemical compound that is extremely soluble in polar solvents and is used in order to obtain lithium metal and as a synthetic reagent. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
CAS Max % 100%
Barium (Ba) 0.003%
Percent Purity 99%
Calcium (Ca) 0.01%
Heavy Metals (as Pb) 0.002%
Insoluble Matter 0.01%
Loss on Drying 1%
Nitrate 0.001%
Packaging Amber Glass Bottle
Potassium (K) 0.01%
Sodium (Na) 0.2%
Sulfate 0.01%
Quantity 2.5 kg
Show More Show Less